Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taurodeoxycholic acid sodium salt ≥95%

Supplier: Ambeed
Warning
Synonyms: Sodium 2-((R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulfonateCyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulfonateCyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulphonateSodium 2-((R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulphonateSodium taurodeoxylateTaurodeoxycholate sodium salt

A108663-5G A108663-100G
77599-572EA 715 USD InStock
77599-572 103823-162
Taurodeoxycholic acid sodium salt ≥95%
Taurodeoxycholic acid sodium salt
Caution: Keep in a dark place, inert atmosphere, room temperature


Formula: C₂₆H₄₄NO₆S·Na
MW: 521.7 g/mol
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00003671
CAS Number: 1180-95-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty