Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-1,3-propanediamine hydrochloride 98%

Supplier: Ambeed
Warning
Synonyms: tert-Butyl (3-aminopropyl)carbamate hydrochloride

A100445-5G A100445-1G A100445-5G A100445-1G A100445-250MG
76717-488EA 10.3 USD InStock
76717-488 77296-686 103821-138 77296-684 103795-442
N-Boc-1,3-propanediamine hydrochloride 98%
N-Boc-1,3-propanediamine hydrochloride
Caution: for Research Use Only


Formula: C₈H₁₉ClN₂O₂
MW: 210.702 g/mol
Boiling Pt: 307.2 °C (760 mm Hg)
Melting Pt: 159…161 °C
Flash Pt: 139.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01076598
CAS Number: 127346-48-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty