Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl-3-aminoazetidine-1-carboxylate 97%

Supplier: Ambeed
Danger
Synonyms: 1-BOC-3-aminoazetidinetert-Butyl 3-aminoazetidine-1-carboxylate3-Amino-1-Boc-azetidine

A162617-25G A162617-5G A162617-1G A162617-10G A162617-1G
76997-862EA 52.53 USD InStock
76997-862 103843-890 77319-950 103805-640 77319-952
tert-Butyl-3-aminoazetidine-1-carboxylate 97%
tert-Butyl-3-aminoazetidine-1-carboxylate
Caution: for Research Use Only


Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
Boiling Pt: 236.6 °C (760 mm Hg)
Flash Pt: 96.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861753
CAS Number: 193269-78-2
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty