Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraphenylmethane 97%

Supplier: Ambeed
Warning

A235754-1G A235754-1G A235754-5G A235754-5G A235754-10G A235754-250MG A235754-25G
103850-796EA 49.62 USD InStock
103850-796 77031-184 77031-182 77089-814 77089-812 103811-028 77304-018
Tetraphenylmethane 97%
Tetraphenylmethane
Caution: for Research Use Only


Formula: C₂₅H₂₀
MW: 320.43 g/mol
Boiling Pt: 431 °C (760 mm Hg)
Melting Pt: 281…286 °C
Flash Pt: 212.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014428
CAS Number: 630-76-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty