Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-N-Boc-1,4-cyclohexanediamine 95%

Supplier: Ambeed
Warning
Synonyms: trans-tert-Butyl (4-aminocyclohexyl)carbamatetrans-4-(Boc-amino)cyclohexylamine

A141840-500G A141840-5G A141840-250MG A141840-25G A141840-5G A141840-100G A141840-1G A141840-10G A141840-1G
76798-340EA 51.5 USD InStock
76798-340 77299-258 76798-342 76798-344 77299-260 103834-106 77316-260 76798-338 103803-070
trans-N-Boc-1,4-cyclohexanediamine 95%
trans-N-Boc-1,4-cyclohexanediamine
Caution: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
Boiling Pt: 322 °C (760 mm Hg)
Flash Pt: 148.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001719
CAS Number: 177906-48-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty