Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylene 98%

Supplier: Ambeed
Danger
Synonyms: 9,10-BenzophenanthreneIsochrysene

A666345-250MG A666345-5G A666345-25G A666345-5G A666345-1G
103840-472EA 10.38 USD InStock
103840-472 76844-674 76844-672 77035-564 76844-676
Triphenylene 98%
Triphenylene
Caution: for Research Use Only


Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 425 °C (760 mm Hg)
Melting Pt: 195…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00001108
CAS Number: 217-59-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty