Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanillic acid 98%

Supplier: Ambeed
Warning
Synonyms: 4-Hydroxy-3-methoxybenzoic acid

A271541-25G A271541-100G A271541-10G A271541-5G
77092-738EA 9.27 USD InStock
77092-738 77250-374 77250-372 103852-962
Vanillic acid 98%
Vanillic acid
Caution: for Research Use Only


Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 353.4 °C (760 mm Hg)
Melting Pt: 208…210 °C
Flash Pt: 149.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty