Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Glu-OtBu 97%

Supplier: Ambeed
Warning
Synonyms: 1-tert-Butyl N-Benzyloxycarbonyl-L-glutamateN-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl esterN-Benzyloxycarbonyl-L-glutamic acid 1-tert-Butyl esterN-Cbz-L-glutamic acid 1-tert-Butyl esterN-Carbobenzoxy-L-glutamic acid 1-tert-Butyl ester1-tert-Butyl N-Cbz-L-glutamateZ-Glu-OtBu1-tert-Butyl N-Carbobenzoxy-L-glutamate

A302765-10G A302765-100G A302765-1G A302765-5G A302765-1G A302765-500G A302765-5G A302765-25G
77114-508EA 868.29 USD InStock
77114-508 77114-504 103835-508 77114-506 103814-348 76838-192 76838-194 77305-856
Z-Glu-OtBu 97%
Z-Glu-OtBu
Caution: for Research Use Only


Formula: C₁₇H₂₃NO₆
MW: 337.372 g/mol
Boiling Pt: 522.6 °C (760 mm Hg)
Flash Pt: 269.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00077016
CAS Number: 5891-45-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty