Would you like to log in again?
V Agar with CNA, Selective for Gardnerella vaginalis, Hardy Diagnostics
Supplier: Hardy Diagnostics
A81
76479-592PK 37.16 USD InStock
76479-592
V Agar with CNA, Selective for Gardnerella vaginalis, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the selective isolation of Gardnerella vaginalis. (Columbia agar base with Colistin, Nalidixic acid, and 5% Human blood).
Ordering information: This product has a short shelf-life.
Delivery information: Ships overnight from California.
Caution: Use biohazard precautions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty