Would you like to log in again?
AnaeroGRO™ Chopped Meat Carbohydrate Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AG20H
76479-598PK 124.52 USD InStock
76479-598
AnaeroGRO™ Chopped Meat Carbohydrate Broth, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
AnaeroGRO™ Chopped Meat Media is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms, especially Clostridium species.
  • Flat bottom contributes to stability when set on end
  • Leak-proof caps provide security during handling and transportation
  • Optically clear tubes are made of clarified glass for clear viewing
  • To avoid oxygen exposure, liquid specimens may be injected directly through the rubber septum of the hungate screw cap with a needle and syringe
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty