Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride solution 5 M, sterile

Supplier: G-Biosciences
Synonyms: SaltHaliteSea saltRock saltSaline

R006 R006
82023-090EA 62.91 USD InStock
82023-090 CA82023-090
Sodium chloride solution 5 M, sterile
Sodium chloride
A sterile solution of 5M sodium chloride prepared with ACS Grade sodium chloride.


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty