Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dodecyl sulfate (SDS) solution 10%

Supplier: G-Biosciences
Warning
Synonyms: Sodium dodecyl sulfateLauryl sulfate sodium saltSodium lauryl sulfateDodecyl sulfate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphateDodecyl sulphate sodium saltDodecyl sulfuric acid sodium salt

R014 R014
82021-474EA 51.54 USD InStock
82021-474 CA82021-474
Sodium dodecyl sulfate (SDS) solution 10%
Sodium dodecyl sulphate (SDS)
Anionic detergent.

  • Critical micelle concentration (CMC): 7-10 mM (25 °C)
  • Aggregation number: 62
  • Average micellar weight: 18,000


Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty