Would you like to log in again?
Thioglycollate with H and K (Hemin and Vitamin K), Hardy Diagnostics
Supplier: Hardy Diagnostics
K22
76377-468PK 51.31 USD InStock
76377-468
Thioglycollate with H and K (Hemin and Vitamin K), Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the cultivation of anaerobic bacteria.
  • Thioglycollate without Indicator with Hemin and Vitamin K
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty