Would you like to log in again?
DTM (Dermatophyte Test Media) Agar Slant, Hardy Diagnostics
Supplier: Hardy Diagnostics
L27
76377-466PK 139.07 USD InStock
76377-466
DTM (Dermatophyte Test Media) Agar Slant, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
(Dermatophyte Test Medium) For the isolation and identification of dermatophytic fungi such as Microsporum canis, the causative agent of ringworm.
  • Vials are made of shatter resistant polycarbonate plastic with a wide mouth opening for easy inoculation
It is also designed to detect dermatophytes such as Epidermophyton and Trichophyton spp. The addition of Gentamicin and Chlortetracycline act as broad spectrum antimicrobics to inhibit a wide range of interfering gram-positive and gram-negative bacteria. Cycloheximide is added to inhibit saprophytic non-pathogenic fungi. Phenol red, the pH indicator, is affected by the presence of dermatophytes (EpidermophytonMicrosporum, and Trichophyton spp.), which all produce alkaline metabolites.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty