Would you like to log in again?
KIMBLE® Heavy-Duty Carboy, DWK Life Sciences
Supplier: DWK Life Sciences
14950-25A
76470-390CS 336.69 USD InStock
76470-390
KIMBLE® Heavy-Duty Carboy, DWK Life Sciences
Bottles Carboys
KIMBLE® carboy-style bottle with sloping shoulders.
  • Neck is tooled for a uniform fit with a no. 12 rubber stopper
  • Size is designed from Federal Specification DD-B-597
  • Non sterile
  • Autoclavable
The heavy-duty construction of these carboys is designed for long-term durability with day to day usage. Ideal for storage and dispensing of solutions. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Accessories include carboy clamp and silicone stopper assemblies. The 3-port, platinum-cured stopper assembly and sanitary PVDF clamp are designed to provide sterile fluid transfer with no extractables, as required in bio-tech and pharmaceutical applications. The stopper ports are designed to connect ¹/₈", ¹/₄" (4-port stopper has two ¹/₄" ports) and ³/₈" silicone tubing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty