Would you like to log in again?
Emmon's Sabdex (Sabouraud Dextrose) Agar, HardyFlask™, Hardy Diagnostics
Supplier: Hardy Diagnostics
X57
76359-400PK 88.06 USD InStock
76359-400
Emmon's Sabdex (Sabouraud Dextrose) Agar, HardyFlask™, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Emmon's Sabdex Agar is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.
  • Lightweight for reduced shipping cost and less chance of breakage
  • Stackable for increased conservation of space, both in incubator and refrigerator. No special storage racks are needed
  • Interlocking for increased stability when incubating and transporting
  • Large surface area for the inoculum
  • Slanted neck facilitates easy inoculation of the flat media surface
  • Improved clarity for better visibility of the growing culture, both front and back
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty