Would you like to log in again?
SARS-CoV-2 Positive Control (N gene), New England Biolabs
Supplier: New England Biolabs
N2117S N2117S
76486-076EA 74 USD InStock
76486-076 CA76486-076
SARS-CoV-2 Positive Control (N gene), New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
The SARS-CoV-2 Positive Control (N gene) is a plasmid that contains the SARS-CoV-2 N-gene (GenBank: MN908947.3). It includes a T7 RNA promoter sequence for generation of RNA transcripts, if necessary. This product is used for assay validation of RT-qPCR, LAMP, or other detection methods for SARS-CoV-2 that target the N gene. The concentration is approximately 1000000 copies/µl.
  • Contains full SARS-CoV-2 nucleocapsid protein (N-gene)
  • Includes T7 RNA promoter sequence for generation of RNA transcripts
  • 1000000 copies/ul concentration
Please note that it is not recommended to use the Positive Control concentration as an absolute copy number for quantitative experiments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty