Would you like to log in again?
Monarch® gDNA Removal Columns, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Monarch®
T2017L
103529-158EA 167 USD InStock
103529-158
Monarch® gDNA Removal Columns, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Monarch gDNA Removal Columns, supplied with the Monarch Total RNA Miniprep Kit, bind genomic DNA during the total RNA miniprep workflow, removing it from your RNA preps. Columns are supplied with Monarch Collection Tubes II , and are compatible with vacuum manifolds.
  • A component of the Monarch Total RNA Miniprep Kit
  • Binds genomic DNA during the total RNA miniprep workflow to remove it from your RNA prep
  • Contain 100 columns and 100 collection tubes
  • Also facilitates reduction of sample viscosity as it passes through
  • Monarch kit components are also available separately for added convenience
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty