Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-Camptothecin 2 mM

Supplier: G-Biosciences
Synonyms: 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione(+)-Camptothecin

API-02 API-02
82021-598EA 162.24 USD InStock
82021-598 CA82021-598
(S)-(+)-Camptothecin 2 mM
(S)-(+)-Camptothecin
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).

  • Supplied as 1ml 2mM

G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.

Camptothecin is a cytotoxic quinoline alkaloid that inhibits the DNA enzyme topoisomerase I (Topo I).


Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
MDL Number: MFCD00081076
CAS Number: 7689-03-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty