Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dexamethasone 100 mM

Supplier: G-Biosciences
Synonyms: Dexamethasonum9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion

API-04 API-04
82021-602EA 159.76 USD InStock
82021-602 CA82021-602
Dexamethasone 100 mM
Dexamethasone
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).

G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.

Dexamethasone is a potent and stable glucocorticoid steroid hormone that is often used as an anti-inflammatory and anti-rheumatic drug.
The mechanism of action is not fully understood, however it is thought that it may inhibit prostaglandin synthesis.


Formula: C₂₂H₂₉FO₅
MW: 392.47 g/mol
MDL Number: MFCD00064136
CAS Number: 50-02-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty