Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Justrite® 5 Gallon Plastic Safety Can with Stainless Steel Piston Pump

Supplier: Justrite
14586
76474-342EA 859.24 USD InStock
76474-342
Justrite® 5 Gallon Plastic Safety Can with Stainless Steel Piston Pump
Pumps Drum/transfer Pumps
A Justrite exclusive, all in one storage and dispensing solution for flammable or combustible liquids.

  • Stainless steel pump
  • Can dispense alcohol based hand sanitizer
  • Self-closing vented filling spout
  • 12" reach below lid

This Type I, FM approved safety can is fitted with a stainless steel pump that allows storing up to 5 gallons of flammable or combustible liquids while also allowing their dispensing at the point of use. The wide filling spout makes it easy to receive liquids from bulk containers and includes a self-closing cap that safely provides pressure relief in the event of a fire. The attached manually operated piston pump delivers up to 3.5 oz. per stroke and a vacuum-relief valve allows liquids to flow smoothly in a controlled and safe manner. The pump body is constructed with stainless steel with PTFE seals, offering superior chemical resistance and a long life. Included internal flash arrestor keeps contents safe from external flame flashbacks. Justrite's unique, conductive carbon insert within the container allows for proper grounding with only one connection point for proper static control. Uniquely ideal for Class 1B and Class 1C alcohol based hand sanitizers.

Certifications: FM approved.

Ordering information: One-year warranty. For chemical compatibility, contact your Avantor representative or customer service.

Delivery information: Includes can with pump.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty