Would you like to log in again?
Axygen® Agarose, PCR Screening, Corning
Supplier: Corning
Axygen®
AGR-MS-50
75816-212CS 235.47 USD InStock
75816-212
Axygen® Agarose, PCR Screening, Corning
Electrophoresis Gels Agarose and Acrylamide
A high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.
  • High resolution of short PCR products and DNA fragments for improved clarity of the gel
  • Stronger gel structure and higher gel strength for better handling
  • The chances of gels breaking or cracking are greatly minimized, even at lower concentrations
  • High gel strength allows for use in blotting
  • Low DNA binding for easy recovery
A molecular screening agarose for improved resolution of DNA fragments with 500 bp or less, ideal for primer-sized fragments.
At a 3% concentration, Agarose MS gives a resolution of DNA fragments similar to gels with polyacrylamide at concentrations of 8%.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty