Would you like to log in again?
Continuous Repositionable Vinyl Cloth Wire and General ID Labels, 3" Core, Brady
Supplier: Brady
THT-92-498-2
76461-008RL 290.83 USD InStock
76461-008
Continuous Repositionable Vinyl Cloth Wire and General ID Labels, 3" Core, Brady
Labels
Permanent or repositionable: once applied, B-498 CleanLift® Series Vinyl Cloth will remain in place until repositioning is required.
  • Repositionable
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Material may be used with the Brady Material Dispenser with Liner Remover for simple labeling tasks. The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 76461-384 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6207 for a different printer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty