Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glossy White 2 mil Polyimide Circuit Board Labels, 3" Core, Brady

Supplier: Brady
THT-47-727-10 THT-2-727-10 THT-43-727-10 THT-103-727-10 THT-11-727-10 THT-71-727-20 THT-46-727-10 THT-45-727-10
76461-122RL 838.05 USD InStock
76461-122 76461-124 76461-130 76246-811 76461-132 76246-441 76461-126 76461-118
Glossy White 2 mil Polyimide Circuit Board Labels, 3" Core, Brady
Labels
High temperature polyimide (B-727) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component pre-process labeling.

  • Heat-resistant, permanent adhesive
  • Service temperature: ‒70 to +100 °C (‒94 to +212 °F)

Ultra-durable adhesive is designed for use with extreme wash protocol and cleaning chemicals, and is ideal for use in auto apply equipment. Glossy white polyimide material is thermal transfer printable.

Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter A8500 Printer Applicator, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.

Certifications: Meets requirements of MIL-STD-202G, Method 215K.

Ordering information: A ribbon is required to print this material. Use Part Number 76244-722 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 76241-622 for a different printer.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty