Would you like to log in again?
High Temperature 12 to 8 Gauge Wire Marking Sleeves, 3" Core, Brady
Supplier: Brady
PermaSleeve®
2HT-250-2-WT-S
76461-296RL 1681.66 USD InStock
76461-296
High Temperature 12 to 8 Gauge Wire Marking Sleeves, 3" Core, Brady
Labels
PermaSleeve® Heat-Shrink Polyvinylidene Fluoride (B-345) wire marker sleeves are specifically designed for high temperature and low vacuum outgassing applications.
  • Flame-retardant, heat-shrinkable
  • Double-side printable
PermaSleeve® 2:1 heat shrinkable sleeves are fade-resistant and flame-retardant, and meet the requirements of NASA Vacuum Outgassing specification SP-R-0022A. Sleeves can be used in applications that require greater resistance to harsh chemicals such as fuels, lubricants, and high power cleaning solvents. The operating temperature range is ‒55 to +225 °C (‒67 to +437 °F).
Sleeves are designed for use with BBP®72 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Certifications: Meets SAE-AMS-DTL-23053/18 (Class 2), SAE-AS-81531, MIL-STD-202, Tested per ASTM E 162, ASTM E 662. RoHS compliant. UL recognized.
Ordering information: A ribbon is required to print this material. Use Part Number 89077-379 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 101076-574 for a different printer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty