Would you like to log in again?
Graduated Pipettes, United Scientific Supplies
Supplier: United Scientific Supplies
UNGRPIP-4ML-PK/500 UNGRPIP-5ML-STPK/500 UNGRPIP-5ML-PK/500 UNGRPIP-4.5ML-PK/500
470334-432PK 55 USD InStock
470334-432 470334-430 470334-428 470334-426
Graduated Pipettes, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Perfect for classroom and laboratory use.
  • General use
  • Graduated
Disposale transfer pipettes are made from low-density polyethylene (LDPE) are ideal for general use. Each pipettes has molded graduations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty