Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disposable Stethoscope, DUKAL™ Corporation

Supplier: Dukal
1115
76473-020CS 218.18 USD InStock
76473-020
Disposable Stethoscope, DUKAL™ Corporation
Stethoscopes
Disposable stethoscope designed for single patient use.

  • Single patient use
  • Overall length: 32"
  • Individually poly bagged

Single head stethoscope is equipped with 22" tubing.

Packaging: Stethoscopes are packaged 10 per box, 10 boxes per case.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty