Would you like to log in again?
Field Sequencing Kit, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
SQK-LRK001
76487-144EA 599 USD InStock
76487-144
Field Sequencing Kit, Oxford Nanopore Technologies
Nucleic Acid Reagents Next Generation Sequencing Reagents
The Field Sequencing Kit delivers cold chain-free nanopore sequencing library preparation for ultimate portability.
  • Cold storage not required — ideal for sequencing in the field
  • Sequence native DNA — eliminate PCR bias and retain base modifications
  • Fast library prep — 10 minutes from extracted DNA
  • Minimal lab equipment required
This kit is recommended for users who are working in the field, with limited access to cold storage and laboratory equipment.
The Field Sequencing Kit generates sequencing libraries from extracted gDNA in 10 minutes using a simple two-step protocol. At the heart of the kit is a transposase which simultaneously cleaves template molecules and attaches tags to the cleaved ends. Rapid Sequencing Adapters are then added to the tagged ends.
The kit is optimized to be stable at ambient temperatures for extended periods, with many of the reagents supplied in lyophilized form. The stability of the components in this kit enables it to be highly portable, making it ideal for use in the field where access to cold storage may be limited.
The workflow is PCR-free and therefore removes PCR bias and retains information about base modifications, which can be analysed using tools developed in the Nanopore Community.
This product includes a flow cell priming kit.
For sample multiplexing see expansion packs.
Delivery information: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location. The kit can be stored at up to 30 °C for one month unopened, or at 2 to 8 °C for three months unopened.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty