Would you like to log in again?
SARS-CoV-2 Spike NTD-coupled Magnetic Beads, Acro Biosystems
Supplier: Acro Biosystems
MBSK01910MG MBSK0192MG
103888-110EA 2692.31 USD InStock
103888-110 103888-112
SARS-CoV-2 Spike NTD-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The SARS-CoV-2 Spike NTD-coupled magnetic beads is produced by coupling biotinylated SARS-CoV-2 spike NTD protein to streptavidin-conjugated magnetic beads. Because Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10 to 14 mol/L, the biotinylated protein can bind to the SA beads irreversibly. The pre-coupled beads are ready to use for capturing anti-SARS-CoV-2 antibody or ACE2 protein from your sample with high specificity.
Antibody Purification in four steps:
1. Resuspend the lyophilized beads by adding the buffer of choice.
2. Add analyte to the suspension, mix and incubate to enable specific binding of the beads and the target protein.
3. Magnetize beads, remove supernatant, and wash unbound protein fractions to capture target protein-bound beads.
4. Wash, magnetize the beads and collect purified target protein for use in downstream applications.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty