Would you like to log in again?
Anti-SARS-CoV-2 (P.1) Antibody IgG Titer Serologic Assay kit (Spike RBD), Acro Biosystems
Supplier: Acro Biosystems
RAST03296TESTS
103888-186EA 700 USD InStock
103888-186
Anti-SARS-CoV-2 (P.1) Antibody IgG Titer Serologic Assay kit (Spike RBD), Acro Biosystems
Assays Immunological Assays
This kit is developed for serologic test for IgG titer of Anti-SARS-CoV-2 Spike RBD antibody in serum/plasma in vitro. The Spike RBD antigen used in this kit contains K417T, E484K, N501Y mutations found in the Brazilian variant (known as P.1).
Multiple variants of SARS-CoV-2 are circulating globally and posting new challenges to human health. The variant P.1 identified in Brazil in Jan, 2021 shows increased transmissibility and is likely to escape from established immunity. There are 12 mutations on the spike protein of the Brazilian variant (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F); of which 3 (K417T, E484K, N501Y) resides on the receptor-binding domain (RBD). To evaluate the impacts of the new variants, a rapid and effective assay kit detecting the levels of antibody against the mutant is in urgent need.
Your experiment will include four simple steps:
a) Add your sample to the plate. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted Secondary antibody HRP-Anti-Human IgG to the plate. The Secondary antibody is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty