Would you like to log in again?
VWR® Standard / VWR® Advanced Standard and Advanced Coaxial Compound Microscopes
Supplier: VWR
VWR®
L790-1 L790-3 L790-2
470331-638EA 511.05 USD InStock
470331-638 470331-642 470331-640
VWR® Standard / VWR® Advanced Standard and Advanced Coaxial Compound Microscopes
Microscopes
High quality performance at an economical price.
  • Wide-field 10× eyepiece with pointer
  • Adjustable brightness
  • Parfocal and parcentered optics
  • Coaxial coarse and fine adjustment controls
  • Three-year manufacturer's warranty
These compound microscopes offer the versatility and precision necessary for research and clinical applications. With all-metal bodies and all-glass-coated optics, they provide crisp, clear images allowing students to view more details and minute structures.
Delivery information: All microscopes include a manual and a dust cover.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty