Would you like to log in again?
Human Serum AB (Off the Clot)
Supplier: Gemini Bio-Products
100-318-100
76683-618EA 443.36 USD InStock
76683-618
Human Serum AB (Off the Clot)
Sera (Serum)
Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
  • Sterile-filtered
  • Ships on dry ice
  • Gamma irradiation available
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty