Would you like to log in again?
GemCell Select™ U.S. Human Serum AB - 16 Donor Max
Supplier: Gemini Bio-Products
GemCell™
100-812G-100 100-812-100
76683-638EA 285.26 USD InStock
76683-638 76683-632
GemCell Select™ U.S. Human Serum AB - 16 Donor Max
Sera (Serum)
GemCell Select™ Human Serum AB is collected from a maximum of 16 healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma. All donor units are tested for viral markers as defined in 21 CFR Part 1271 and found to be non-reactive.
  • Ideal for clinical applications where small donor pools are necessary due to regulatory requirements
  • 0.1 µm sterile-filtered
  • Gamma irradiation available
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty