Would you like to log in again?
GemCell Plus™ U.S. Human Serum AB
Supplier: Gemini Bio-Products
GemCell™
100-612-100 100-612G-100
76683-628EA 430.76 USD InStock
76683-628 76683-620
GemCell Plus™ U.S. Human Serum AB
Sera (Serum)
GemCell Plus™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma AB. Our GemCell Plus™ goes through additional rigorous viral testing to include West Nile Virus, Chagas, HTLV I/II, and HBc as well as the standard FDA required panel. All donor units are tested for viral markers and found to be non-reactive.
  • 0.1 µm sterile-filtered
  • Gamma irradiation available
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty