Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Recombinant USER® III
Supplier: New England Biolabs
M5509S M5509S
76640-798EA 104 USD InStock
CA76640-798 76640-798
Recombinant USER® III
Enzymes
Thermostable USER (Uracil-Specific Excision Reagent) III Enzyme generates a single nucleotide gap at the location of a uracil residue. It is active between 50-75°C (optimal activity observed at 65°C).
  • Thermostable
  • Isolated from a recombinant source
  • Supplied with 10X Reaction Buffer
Thermostable USER (Uracil-Specific Excision Reagent) III Enzyme generates a single nucleotide gap at the location of a uracil. Thermostable USER III Enzyme is a mixture of Afu Uracil DNA glycosylase (UDG) and the DNA glycosylase-lyase Endonuclease IV. Afu UDG catalyzes the excision of a uracil base, forming an abasic (apyrimidinic) site while leaving the phosphodiester backbone intact. The lyase activity of Endonuclease IV breaks the phosphodiester backbone at the 3’ and 5’ sides of the abasic site so that base-free deoxyribose is released leaving a 5’ deoxyribose phosphate (dRP) and a 3’-hydroxyl. Thermostable USER III Enzyme also has 3’-diesterase activity which can remove 3’ phosphate, 3’-alpha, beta-unsaturated aldehyde, phosphoglycoaldehyde, and other 3' blocking groups. Thermostable USER III Enzyme is optimally active at 65 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty