Would you like to log in again?
APO-10
Supplier: EMD Millipore
178375-1GM 178375-1GM
80057-804EA 171.55 USD InStock
80057-804 CA80057-804
APO-10
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Novel non-ionic detergent useful for plasmid DNA isolation. Plasmid DNA isolated using APO-10 does not require the use of phenol/chloroform or proprietary resins. Provides excellent yields of high quality DNA.
  • CAS No.: 2190-95-6
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty