Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HardyCHROM™ Campy Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G339 G339
76595-432PK 74.72 USD InStock
CA76595-432 76595-432
HardyCHROM™ Campy Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Chromogenic media to detect Campylobacter spp. with color read out in 48 hours or less!
  • Red colonies stand out on a translucent background making detection easy.
  • Highly selective for Campylobacter species from mixed samples, especially C. jejuni, C. lari, C. coli, and C. fetus.
  • Flexible workflow can be used with existing FDA-BAM or USDA-FSIS pre-enrichment procedures, or for direct plating procedures.
  • Saves time and money by eliminating needless work-up of suspicious colonies.
HardyCHROM Campy Agar is recommended as a screening medium for the slective isolation and chromagenic differentiation of Campylobacter spp. from direct stool cultures or from food or poultry samples. Some ingrediens are very light sensitive. Store and incubate media in the dark.
Packaging: 10 per pack
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty