Would you like to log in again?
Peacock™ Prestained Protein Markers, Biotium
Supplier: Biotium
21530-50UL 21530-500UL
76221-772EA 72.01 USD InStock
76221-772 76221-606
Peacock™ Prestained Protein Markers, Biotium
Electrophoresis Reagents Molecular Weight Markers
A visible three-color protein marker for SDS-PAGE or western blotting, with 10 bands ranging from 10 to 180 kDa.
  • Three-color visible protein ladder
  • 10 total protein bands, ranging from 10 kDa
  • Eight blue bands, plus a red band at 75 kDa and green band at 25 kDa
  • Ready-to-load: no heating required
  • Store at −20 °C
Peacock™ Prestained Protein Marker is a three-color protein ladder that allows you to visually monitor protein separation during SDS-PAGE or protein transfer to membranes for western blotting. Biotium also offers Peacock™ Plus Prestained Protein Marker (76221-776,76221-774), a three-color marker with 12 bands ranging from 8 to 245 kDa.
The Peacock™ Prestained Marker contains a total of 10 visible bands. This includes 8 blue bands ranging from 10 to 180 kDa, plus a red band at 75 kDa and green band at 25 kDa for easy band identification. Peacock™ Prestained Protein Marker is ready to load with no heating or other preparation needed. Recommended loading is 3 to 5 µl per well for mini gels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty