Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ampicillin sodium salt, Lab-Grade

Supplier: ALDON CORP SE
Danger
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

AA0356-5G
470024-776EA 61.3 USD InStock
470024-776
Ampicillin sodium salt, Lab-Grade
Ampicillin sodium salt


Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty