Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Borosilicate Glass Test Tubes, 25 ml
Supplier: EISCO SCIENTIFIC LLC
PH0941BGTT
470335-372PK 10.6 USD InStock
470335-372
Borosilicate Glass Test Tubes, 25 ml
Tubes Test Tubes
A pair of tubes for elecrolysis experiments.
  • 25 ml capacity each
  • Scale printed inverse
This pair of test tubes is most commonly used for electrolysis experiments which require an upside-down test tube to capture gas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty