Would you like to log in again?
SaniPath Disinfecting Wipes, Mortech®
Supplier: Mortech
BE036
76498-452EA 62.41 USD InStock
76498-452
SaniPath Disinfecting Wipes, Mortech®
Wipes Disposable Wipes and Towels Disinfectant Wipes
SaniPath wipes are a one-step cleaning and disinfecting necessity for all pathology/necropsy lab surfaces.
  • 160 premoistened towelettes per pop-up canister
  • Bleach-free and phenol-free
  • EPA approved; EPA Reg. No. 1839-174-74952, EPA Est. No. 5747-MI-01
The wipes allow users to clean and disinfect quickly, with or without gloves. They’re effective against a wide range of pathogenic bacteria, viruses, and fungi. This product is ideal for lab telephones, computers, keyboards, writing instruments, splash shields, etc. Set these next to pathology photography stations to remove blood spots and smears. They also come in handy when cleaning pathology lab refrigerators and freezer surfaces. Use daily for a cleaner safer work environment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty