Would you like to log in again?
Economy Hospital Body Bag, with Envelope Zipper, Mortech®
Supplier: Mortech
BE082
76498-504EA 65.32 USD InStock
76498-504
Economy Hospital Body Bag, with Envelope Zipper, Mortech®
Bags Emergency Bags
Economy hospital body bag.
  • Overeall dimensions are 36W×90"L
  • Envelope-style, rust-resistant zipper, with two pulls
  • Without handles
  • 10 year shelf life
Economy body bags are constructed from 5.5 mil high-strength, white, virgin PVC material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty