Would you like to log in again?
Acrylic Bowl, Mortech®
Supplier: Mortech
BB024
76498-254EA 130.12 USD InStock
76498-254
Acrylic Bowl, Mortech®
Trays/Dishes/Pots Laboratory Trays/Dishes
Multi-purpose, washable acrylic bowl for isolating or weighing specimens.
  • 5.25" base Ø
  • 6.25" top Ø
  • Plastic construction
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty