Would you like to log in again?
SaniPath Disinfecting Spray Cleaner, Mortech®
Supplier: Mortech
BE047
76498-466EA 33.39 USD InStock
76498-466
SaniPath Disinfecting Spray Cleaner, Mortech®
Cleaning Agents Disinfectants
Protect personnel and their work areas with SaniPath Disinfecting Spray, a multi-purpose decontaminating cleaner. It is a bactericidal, fungicidal, virucidal, and Tuberculocide.
  • Mild, lemon fragrance leaves the lab smelling fresh
  • 32 oz. container
This cleaner is great for spill clean up and should be used daily for pathology and morgue work surfaces including equipment, tables, coolers, cutting stations, instrumentation, floors, computers, and telephones.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty