Would you like to log in again?
Sulfo-Cyanine 3 NHS ester
Supplier: AAT BIOQUEST INC
264 263
76484-614EA 604.2 USD InStock
76484-614 76484-616
Sulfo-Cyanine 3 NHS ester
Sulfo-Cyanine 3 NHS ester
A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • pH Tolerance From 3-10
  • Bright Fluorescence
  • Multiple Bioconjugation Applications
Caution: For Research Use Only
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty