Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfo-Cyanine 5 NHS ester ≥90%

Supplier: AAT BIOQUEST INC

268 269
76484-622EA 604.2 USD InStock
76484-622 76484-624
Sulfo-Cyanine 5 NHS ester ≥90%
Sulfo-Cyanine 5 NHS ester
A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.

  • pH Tolerance From 3-10
  • Bright Fluorescence
  • Multiple Bioconjugation Applications

Caution: For Research Use Only


CAS Number: 1448789-45-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/11/94/43921194.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty