Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide silylation reagent, Pierce™

Supplier: Thermo Scientific
Synonyms: Methyl sulfoxideDMSOMethyl sulphoxide

TS-20684
10066-276EA 90.57 USD InStock
10066-276
Dimethyl sulfoxide silylation reagent, Pierce™
Dimethyl sulphoxide
Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions. They are purified, dried, and packaged under nitrogen in convenient Thermo Scientific™ Hypo-Vial™ Sample Storage Vials, and are supplied with elastomer septa to enable immediate access to the sample without exposure to moisture and oxygen. Use polar solvents (acetonitrile, dimethylformamide, dimethylsulfoxide, pyridine, tetrahyrdrofuran) to facilitate reactions; nonpolar organic solvents may be used but will not accelerate the rate of reaction.


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
MDL Number: MFCD00002089
CAS Number: 67-68-5
UN: 2265
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty