Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nile Blue A ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: Basic Blue 12Nile blue A sulfateNile blue sulphateNile blue A sulphateNile blue sulfate

60
76480-498EA 93.54 USD InStock
76480-498
Nile Blue A ≥95%
Nile blue A
Nile Blue A is certified for use in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.


Formula: C₄₀H₄₀N₆O₆S
MW: 732.84 g/mol
MDL Number: MFCD00064529
CAS Number: 3625-57-8
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty