Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhodamine 6G ≥90%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

70
76480-512EA 93.54 USD InStock
76480-512
Rhodamine 6G ≥90%
Rhodamine 6G
Rhodamine 6G is also called Rhodamine 590, R6G, Basic Rhodamine Yellow, or C.I.


Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
MDL Number: MFCD00012665
CAS Number: 989-38-8
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty