Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol purple ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST INC
Warning
Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide5',5''-Dibromo-o-cresolsulphonphthalein5',5''-Dibromo-o-cresolsulfonphthalein

92
76480-538EA 361.89 USD InStock
76480-538
Bromocresol purple ≥95%, Ultra Pure Grade
Bromocresol purple
Bromocresol purple (BCP) or 5',5-dibromo-o-cresolsulfophthalein, is a pH indicator.


Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
MDL Number: MFCD00011681
CAS Number: 115-40-2
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty